missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoprotegerin/TNFRSF11B Antibody (98A1071), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56505PCP
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry.
Specifications
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| PerCP | |
| O00300 | |
| TNFRSF11B | |
| A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody. | |
| 0.1 mL | |
| Primary | |
| The Osteoprotegerin antibody, clone 98A1071, detects all forms of Osteoprotegerin including monomeric, dimeric and Osteoprotegerin bound to RANKL (Vandooren, 2008) as well as Osteoprotegerin bound to TRAIL (Sandra, 2006). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 98A1071 | |
| Flow Cytometry 1 μL/1 million cells | |
| OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4982 | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction