missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Osteoprotegerin/TNFRSF11B Antibody (SR2339), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27863-50ul
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| SR2339 | |
| Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| A synthesized peptide derived from human Osteoprotegerin/TNFRSF11B (Uniprot #: O00300) | |
| 50 μL | |
| Apoptosis, Biologically Active Proteins, Cell Biology | |
| 4982 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction