missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osterix/Sp7 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98576AF750
This item is not returnable.
View return policy
Description
Osterix/Sp7 Polyclonal antibody specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot
Specifications
| Osterix/Sp7 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 121340 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| MGC126598, OSXosterix, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix | |
| Produced in rabbits immunized with E. coli-derived Human Osterix/Sp7 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction