missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTF1 Antibody (9G4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59396PCP
This item is not returnable.
View return policy
Description
OSTF1 Monoclonal antibody specifically detects OSTF1 in Human samples. It is validated for Flow Cytometry
Specifications
| OSTF1 | |
| Monoclonal | |
| PerCP | |
| FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1 | |
| Recombinant human OSTF1 (1-217 aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 26578 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 9G4 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction