missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10671-1-AP-20UL
This item is not returnable.
View return policy
Description
Osteoclast-stimulating factor-1 is an intracellular protein produced by osteoclasts that indirectly induces osteoclast formation and bone resorption.Specifications
| OSTF1 | |
| Polyclonal | |
| Unconjugated | |
| OSTF1 | |
| OSF, OSTF1, SH3P2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20409, 259275, 26578 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q62422, Q6P686, Q92882 | |
| Ostf1 | |
| OSTF1 Fusion Protein Ag1072 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction