missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTM1 Antibody (30), DyLight 680 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89708FR
This item is not returnable.
View return policy
Description
OSTM1 Monoclonal antibody specifically detects OSTM1 in Human samples. It is validated for ELISA
Specifications
| OSTM1 | |
| Monoclonal | |
| DyLight 680 | |
| GIPN, GLGAIP-interacting protein N terminus, grey-lethal osteopetrosis, HSPC019, OPTB5, osteopetrosis associated transmembrane protein 1, osteopetrosis-associated transmembrane protein 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human OSTM1 extracellular domain (NP_054747.2; Met 1-Pro 284). | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 28962 | |
| Store at 4&drg;C in the dark. | |
| IgG |
| ELISA | |
| 30 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction