missing translation for 'onlineSavingsMsg'
Learn More

OSU-T315, MedChemExpress

Product Code. 16400058
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400058

Brand: Medchem Express HY18676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OSU-T315 (ILK-IN-1) is a small Integrin-linked kinase (ILK) inhibitor with an IC50 of 0.6 μM, inhibiting PI3K/AKT signaling by dephosphorylation of AKT-Ser473 and other ILK targets (GSK-3β and myosin light chain). OSU-T315 abrogates AKT activation by impeding AKT localization in lipid rafts and triggers caspase-dependent apoptosis in an ILK-independent manner. OSU-T315 causes cell death through apoptosis and autophagy.

OSU-T315 (ILK-IN-1) is a small Integrin-linked kinase (ILK) inhibitor with an IC50 of 0.6 μM, inhibiting PI3K/AKT signaling by dephosphorylation of AKT-Ser473 and other ILK targets (GSK-3β and myosin light chain). OSU-T315 abrogates AKT activation by impeding AKT localization in lipid rafts and triggers caspase-dependent apoptosis in an ILK-independent manner. OSU-T315 causes cell death through apoptosis and autophagy.

Specifications

Chemical Name or Material OSU-T315
Physical Form Solid
CAS 2070015-22-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H30F3N5O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 260 mg/mL (487.27 mM)
SMILES O=C(NC)CCC1=CC(C2=CC=C(C3=CC=C(C(F)(F)F)C=C3)C=C2)=NN1C4=CC=C(N5CCNCC5)C=C4
Molecular Weight (g/mol) 533.59
Formula Weight 533.59
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.