Learn More
OTC Monoclonal Antibody (OTI3H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802533
Encompass_Preferred
 View more versions of this product
Product Code. 17581143
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
Specifications
Specifications
OTC
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P00480
OTC
Human recombit protein fragment corresponding to amino acids 33-354 of human OTC produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI3H4
Unconjugated
OTC
OCTD
Mouse
Affinity Chromatography
RUO
5009
-20° C, Avoid Freeze/Thaw Cycles