missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTC Monoclonal Antibody (OTI4D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802599S
This item is not returnable.
View return policy
Description
OTC Monoclonal antibody specifically detects OTC in Human samples. It is validated for Western BlotSpecifications
| OTC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00480 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5009 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI4D6 | |
| Unconjugated | |
| OTC | |
| AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf | |
| Human recombinant protein fragment corresponding to amino acids 33-354 of human OTC produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction