Learn More
NCF4 Monoclonal Antibody (OTI6H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809224S
Encompass_Preferred
 View more versions of this product
Product Code. 17571234
  • € 242.00 / 30µL
Description
NCF4 Monoclonal antibody specifically detects NCF4 in Human samples. It is validated for Western Blot
Specifications
Specifications
NCF4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15080
Mouse
Affinity chromatography
RUO
4689
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI6H9
Unconjugated
Ncf4
AI451400; CGD3; CTA-833B7.1; MGC3810; NCF; Ncf4; NCF-4; Neutrophil cytosol factor 4; neutrophil cytosolic factor 4; neutrophil cytosolic factor 4, 40kDa; neutrophil NADPH oxidase factor 4; OTTHUMP00000028736; OTTHUMP00000043756; P40PHOX; p40-phox; SH3 and PX domain-containing protein 4; SH3PXD4
Full length human recombinant protein of human NCF4 produced in HEK293T cell.