Learn More
OTC Mouse anti-Human, Clone: OTI4G8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802596
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.Specifications
| OTC | |
| Monoclonal | |
| Unconjugated | |
| OTC | |
| OCTD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5009 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P00480 | |
| OTC | |
| Human recombit protein fragment corresponding to amino acids 33-354 of human OTC produced in SF9 cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.