missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTC Monoclonal Antibody (OTI5H8), TrueMAB™, OriGene
Description
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
Specifications
Specifications
| Antigen | OTC |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5H8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | OTC |
| Gene Accession No. | P00480 |
| Gene Alias | OCTD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?