missing translation for 'onlineSavingsMsg'
Learn More

OTOP2 Polyclonal Antibody

Product Code. 15790512 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15790512 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790512 Supplier Thermo Scientific Supplier No. PA562727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

OTOP2 Polyclonal Antibody

OTOP2 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen OTOP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTOP2
Gene Accession No. Q7RTS6
Gene Alias OTOP2; otopetrin 2; otopetrin-2; Proton channel OTOP2
Gene Symbols OTOP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OTOP2. Recombinant protein control fragment (Product #RP-93515).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.