Learn More
Invitrogen™ OTOS Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA569880
Product Code. 15808293
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Pig: 79%; Rat: 93%.
OTOS gene ontology annotations related to this gene include sensory perception of sound.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OTOS
Polyclonal
Unconjugated
OTOS
Ocp10; Organ of Corti 10 kDa protein; Otos; OTOSP; Otospiralin
Rabbit
Affinity Chromatography
RUO
150677
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8NHW6
OTOS
synthetic peptide directed towards the N terminal region of human OTOS.