Learn More
OTSSP167 hydrochloride, MedChemExpress
Product Code. 16462677
Encompass_Preferred
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Product Code. 16462677
Brand: Medchem Express HY15512A
MedChemExpress OTSSP167 (OTS167) hydrochloride is a highly potent and ATP-competitive MELK inhibitor with IC50 value of 0.41 nM.
OTSSP167 (OTS167) hydrochloride is a highly potent and ATP-competitive MELK inhibitor with IC50 value of 0.41 nM.
Specifications
Chemical Name or Material OTSSP167 hydrochloride
CAS 1431698-10-0
Color Gray
Purity Grade Notes Research
Molecular Formula C25H29Cl3N4O2
Quantity 50 mg
Synonym OTS167 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (63.62 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (13.63 mM; Need ultrasonic)
SMILES ClC1=CC(C2=CC=C(N=CC(C(C)=O)=C3N[C@H]4CC[C@H](CN(C)C)CC4)C3=N2)=CC(Cl)=C1O.[H]Cl
Molecular Weight (g/mol) 523.88
Formula Weight 523.88
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only