missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUB2 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501943S
This item is not returnable.
View return policy
Description
OTUB2 Monoclonal antibody specifically detects OTUB2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| OTUB2 | |
| Monoclonal | |
| 0.78 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96DC9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 78990 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2B2 | |
| Unconjugated | |
| OTUB2 | |
| 2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2 | |
| Full length human recombinant protein of human OTUB2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction