missing translation for 'onlineSavingsMsg'
Learn More

OTUB2 Monoclonal Antibody (OTI6F7), TrueMAB™, OriGene

Product Code. 17555571
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17555571 30 μL 30µL
17535571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17555571 Supplier Origene Technologies Supplier No. TA501942S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OTUB2 Monoclonal antibody specifically detects OTUB2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen OTUB2
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6F7
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OTUB2
Gene Accession No. Q96DC9
Gene Alias 2010015L18Rik; 4930586I02Rik; AI413508; AW557219; C14orf137; deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTB2; OTU deubiquitinase, ubiquitin aldehyde binding 2; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; OTU2; OTUB2; Otubain-2; ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; ubiquitin-specific-processing protease OTUB2
Host Species Mouse
Immunogen Full length human recombinant protein of human OTUB2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78990
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.