missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99440AF700
This item is not returnable.
View return policy
Description
Otubain-1 Polyclonal antibody specifically detects Otubain-1 in Mouse samples. It is validated for Western Blot
Specifications
| Otubain-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Otubain-1 | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 55611 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction