missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-49934
This item is not returnable.
View return policy
Description
Otubain-1 Polyclonal specifically detects Otubain-1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Otubain-1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| OTUB1 | |
| The immunogen for this product maps to a region between residue 1 and 40 of human OTU Domain, Ubiquitin Aldehyde Binding 1 using the numbering given in entry NP_060140.2 (GeneID 55611). | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 55611 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, European domestic ferret, African elephant, Small-eared galago, Thirteen-lined ground squirrel and Little brown bat. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction