missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD1 Polyclonal Antibody

OTUD1 Polyclonal Antibody
Brand: Invitrogen PA565212
This item is not returnable.
View return policy
Description
OTUD1 Polyclonal Antibody for ICC/IF
Specifications
| OTUD1 | |
| Polyclonal | |
| Unconjugated | |
| OTUD1 | |
| DUBA7; DUBA-7; OTDC1; OTU deubiquitinase 1; OTU domain containing 1; OTU domain-containing protein 1; OTUD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 220213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VV17 | |
| OTUD1 | |
| Recombinant Human OTUD1. Recombinant protein control fragment (Product #RP-105951). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction