Learn More
OTUD4 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-37689AF750
Product Code. 18646339
  • € 633.00 / 0.10mL
Description
OTUD4 Polyclonal antibody specifically detects OTUD4 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OTUD4
Polyclonal
DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4
Western Blot
50mM Sodium Borate
Store at 4°C in the dark.