missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00
Specifications
| Antigen | OTUD4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.25-0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
OTUD4 Polyclonal specifically detects OTUD4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
| OTUD4 | |
| Western Blot 0.25-0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| NP_001096123 | |
| 54726 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human OTUD4. The immunogen is located within the last 50 amino acids of OTUD4. Amino Acid Squence: SWKGQPSRSRDEGYQYHR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 125 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4 | |
| OTUD4 | |
| IgG | |
| Affinity Purified | |
| Multiple isoforms of OTUD4 are known to exist. OTUD4 antibody is predicted to not cross-react with other members of the OTUD family. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title