missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-37690CL1
This item is not returnable.
View return policy
Description
OTUD4 Polyclonal antibody specifically detects OTUD4 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| OTUD4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4 | |
| This antibody maps to a region between residue 1063 to 1113 of human OTU Domain Dontaining 4 using the numbering given in entry XP_005263137.1 (GeneID 54726). | |
| 0.1 mL | |
| Cancer | |
| 54726 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction