missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77003MFV610
This item is not returnable.
View return policy
Description
OTUD4 Polyclonal antibody specifically detects OTUD4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| OTUD4 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human OTUD4. The immunogen is located within the last 50 amino acids of OTUD4. Amino Acid Squence: SWKGQPSRSRDEGYQYHR | |
| 0.1 mL | |
| Cancer | |
| 54726 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction