missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35511-20ul
This item is not returnable.
View return policy
Description
OTUD4 Polyclonal antibody specifically detects OTUD4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| OTUD4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| DKFZp434I0721, HIV-1 induced protein HIN-1, HIV-1-induced protein HIN-1, HSHIN1, KIAA1046HIN1DUBA6, OTU domain containing 4, OTU domain-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-500 of human OTUD4 (NP_001096123.1).,, Sequence:, LKSKQAQQKRDYSIAAGLQYEVGDKCQVRLDHNGKFLNADVQGIHSENGPVLVEELGKKHTSKNLKAPPPESWNTVSGKKMKKPSTSGQNFHSDVDYRGPKNPSKPIKAPSALPPRLQHPSGVRQHAFSSHSSGSQSQKFSSEHKNLSRTPSQIIRKPDRERVEDFDHTSRESNYFGLSPEERREKQAIEESRLLYEIQNRDEQAFPALSSSSVNQSASQSSNPCVQRKSSHVGDRKGSRRRMDTEERKDKDSIHGHSQLDKRPEPSTLENITDDKYATVSSPSKSKKLECPSPAEQKPAE | |
| 20 μL | |
| Cancer | |
| 54726 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction