missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OTUD6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598732
This item is not returnable.
View return policy
Description
OTUD6B Polyclonal Antibody for Western Blot, IHC (P), ELISA
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
Specifications
| OTUD6B | |
| Polyclonal | |
| Unconjugated | |
| OTUD6B | |
| 2600013N14Rik; AU015433; CGI-77; Deubiquitinase OTUD6B; DUBA5; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; Otud6b; RGD1310024 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51633 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N6M0 | |
| OTUD6B | |
| Recombinant Human Deubiquitinase OTUD6B protein (1-293AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction