missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD7B/Cezanne/ZA20D1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97842AF594
This item is not returnable.
View return policy
Description
OTUD7B/Cezanne/ZA20D1 Polyclonal specifically detects OTUD7B/Cezanne/ZA20D1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| OTUD7B/Cezanne/ZA20D1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| cellular zinc finger NF-kappaB Cezanne, Cellular zinc finger NF-kappa-B protein, CEZANNEA20 domain containing 1, EC 3.4.19.12, OTU domain containing 7B, OTU domain-containing protein 7B, ZA20D1, Zinc finger A20 domain-containing protein 1, Zinc finger protein Cezanne | |
| Produced in rabbits immunized with E. coli-derived Human OTUD7B/Cezanne/ZA20D1 fragment. | |
| 0.1 mL | |
| Mucosal Immunology | |
| 56957 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction