missing translation for 'onlineSavingsMsg'
Learn More

OTUD7B Monoclonal Antibody (OTI3D10), TrueMAB™, OriGene

Product Code. 17571834
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17571834 30 μL 30µL
17551834 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17571834 Supplier Origene Technologies Supplier No. TA809517S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OTUD7B Monoclonal antibody specifically detects OTUD7B in Human samples. It is validated for Western Blot

Specifications

Antigen OTUD7B
Applications Western Blot
Classification Monoclonal
Clone OTI3D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene WDR92
Gene Accession No. Q96MX6
Gene Alias 2900060B22Rik; 4930463P07Rik; AI462125; cellular zinc finger anti-NF-kappaB Cezanne; cellular zinc finger anti-NF-kappa-B protein; CEZANNE; OTU deubiquitinase 7B; OTU domain containing 7B; OTU domain-containing protein 7B; OTUD7B; RGD1308111; Trabid2; TRAF binding domain containing 2; Za20d1; Zinc finger A20 domain-containing protein 1; zinc finger protein Cezanne; zinc finger, A20 domain containing 1
Host Species Mouse
Immunogen Full length human recombinant protein of human WDR92 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116143
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.