missing translation for 'onlineSavingsMsg'
Learn More

OTUD7B Monoclonal Antibody (OTI3D10), TrueMAB™, OriGene

Product Code. 17551834
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17551834 100 μL 100µL
17571834 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17551834 Supplier Origene Technologies Supplier No. TA809517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Negative regulator of the non-canonical NF-kappa-B pathway that acts by mediating deubiquitination of TRAF3, an inhibitor of the NF-kappa-B pathway, thereby acting as a negative regulator of B-cell responses. In response to non-canonical NF- kappa-B stimuli, deubiquitinates ′Lys-48′-linked polyubiquitin chains of TRAF3, preventing TRAF3 proteolysis and over-activation of non-canonical NF-kappa-B. Negatively regulates mucosal immunity against infections. Mediates deubiquitination of EGFR. Has deubiquitinating activity toward ′Lys-11′, ′Lys-48′ or ′Lys-63′- linked polyubiquitin chains. In vitro, has preference for ′Lys- 11′-linked polyubiquitin chains; however such data are unsure in vivo. Hydrolyzes both linear and branched forms of polyubiquitin.

Specifications

Antigen OTUD7B
Applications Western Blot
Classification Monoclonal
Clone OTI3D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene OTUD7B
Gene Accession No. Q6GQQ9
Gene Alias CEZANNE, ZA20D1
Gene Symbols OTUD7B
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 449-729 of human OTUD7B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56957
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.