missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otx1 Antibody (S04-1D1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19902-50ul
This item is not returnable.
View return policy
Description
Otx1 Monoclonal antibody specifically detects Otx1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Otx1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-1D1 | |
| Unconjugated | |
| FLJ38361, homeobox protein OTX1, MGC15736, orthodenticle (Drosophila) homolog 1, orthodenticle homeobox 1, Orthodenticle homolog 1, orthodenticle homolog 1 (Drosophila) | |
| A synthetic peptide of human Otx1 (Uniprot # P32242) | |
| 50 μg | |
| Neuroscience | |
| 5013 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction