missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTX2 Monoclonal Antibody (OTI3G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809627S
This item is not returnable.
View return policy
Description
OTX2 Monoclonal antibody specifically detects OTX2 in Human samples. It is validated for Western BlotSpecifications
| OTX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9P2E9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6238 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3G9 | |
| Unconjugated | |
| RRBP1 | |
| CPHD6; E130306E05Rik; homeobox Otx2; homeobox protein OTX2; MCOPS5; orthodenticle homeobox 2; orthodenticle homolog 2; OTX; OTX2 | |
| Human recombinant protein fragment corresponding to amino acids 484-590 of human RRBP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction