missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13497-1-AP-20UL
This item is not returnable.
View return policy
Description
OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The proteinmay bind to BCD target sequence (BTS): 5-′TCTAATCCC-3′. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).Specifications
| OTX2 | |
| Polyclonal | |
| Unconjugated | |
| OTX2 | |
| Homeobox protein OTX2, MCOPS5, orthodenticle homeobox 2, Orthodenticle homolog 2, OTX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18424, 305858, 5015 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32243, P80206, Q64201 | |
| Otx2 | |
| OTX2 Fusion Protein Ag4323 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction