missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (MIC1-1C3), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-18961H
This item is not returnable.
View return policy
Description
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Specifications
| Oval Cell Marker | |
| Monoclonal | |
| HRP | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MIC1-1C3 | |
| PBS | |
| Nonparenchymal cells from DDC-treated mice | |
| 0.1 mL | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction