missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (OC2-2F3), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-18964MFV610
This item is not returnable.
View return policy
Description
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry
Specifications
| Oval Cell Marker | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Nonparenchymal cells from DDC-treated mice | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OC2-2F3 | |
| Flow Cytometry | |
| Rat | |
| Tissue culture supernatant | |
| RUO | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction