missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (OC2-4E8), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-18971IR
This item is not returnable.
View return policy
Description
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Oval Cell Marker | |
| Monoclonal | |
| DyLight 755 | |
| Rat | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| OC2-4E8 | |
| 50mM Sodium Borate | |
| Nonparenchymal cells from DDC-treated mice to induce the oval cell response. | |
| 0.1 mL | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction