Learn More
Brady™ Oval Polyethylene Tags
Tag a variety of assets with this tough polyethylene tag label material (B-145) that prints on both sides and folds to form an oval tag. Easily print anywhere with a Brady™ handheld label printer.
Brand:  Brady™ 170851
Additional Details : Weight : 0.17700kg
Product Code. 17851769
  • € 81.69 / Pack of 65
Estimated Shipment: 26-06-2024
to see stock.
Description
Description
  • Suitable for a wide range of applications including lockout and safety warning repair
  • Designed to resist tearing and exposure to weather and extreme temperatures
  • Simply print, fold, and easily attach using cable ties
Specifications
Specifications
RoHS Compliant to 2005/618/EC MCV Amendment to RoHS Directive 2002/95/EC
Black on Gray
Die-cut
Labels
Asset Tracking, Component and Equipment Labeling, Data and Telecommunications Labeling, Facility Identification, General Identification, Inventory and Inspection Labeling, Safety Identification, Wire and Cable Labeling
65 Tags/Cartridge
0.023 in.
0.75 in.
3.1 cm
Non-adhesive
Matte
Polyethylene
Non-sterile
BMP51, M511
-40°F to +212°F
0.75 x 1.206 in.
1.206 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers