Learn More
VistaLab™ Technologies Ovation Base Retainer Screw
Maintenance part
Brand: VistaLab™ Technologies 9057-4006
Additional Details : Weight : 0.10000kg
Description
Base retainer screw with O-Ring for Ovation pipettesSpecifications
Non-sterile | |
O-Ring | |
2 in. | |
1 in. |
Ovation QS, F1, F2, E8, E12, Macro5 & Macro10 | |
1 Ea. | |
1 in. |