Learn More
VistaLab™ Technologies Ovation Base Retainer Screw
Maintenance part
Brand:  VistaLab™ Technologies 9060-4006
Additional Details : Weight : 0.43090kg
 View more versions of this product
Product Code. 16485204
  • € 18.38 / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
Base retainer screw with O-Ring for Ovation pipettes
Specifications
Specifications
Non-sterile
O-Ring
5 in.
1 in.
Ovation ESC
1 Ea.
3 in.