missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVGP1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76939UV
This item is not returnable.
View return policy
Description
OVGP1 Polyclonal antibody specifically detects OVGP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| OVGP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 5016 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| CHIT5, EGP, Estrogen-dependent oviduct protein, MUC9, mucin 9, mucin-9, OGP, oviduct glycoprotein, Oviductal glycoprotein, oviductal glycoprotein 1, 120kDa, oviductin, oviduct-specific glycoprotein | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human OVGP1. The immunogen is located within amino acids 60 - 110 of OVGP1. Amino Acid Squence: ILYPEFNKLKERNRELKT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction