missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OVGP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520580
This item is not returnable.
View return policy
Description
Despite its predicted molecular weight, OVGP1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human placenta lysate. PA5-20580 can be used with blocking peptide PEP-0700.
Oviductins belong to a family of glycoproteins that have been suggested to play several roles during the early processes of reproduction. OVGP1 is a large carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites within threonine, serine, and proline-rich tandem repeats. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. At least two isoforms of OVGP are known to exist.
Specifications
| OVGP1 | |
| Polyclonal | |
| Unconjugated | |
| OVGP1 | |
| 120kDa; AU016433; AU019448; CHIT5; chitinase 5; EGP; Estrogen-dependent oviduct protein; MOGP; MUC9; mucin 9; mucin-9; OGP; Ovgp1; oviduct glycoprotein; oviduct glycoprotein OGP; Oviductal glycoprotein; oviductal glycoprotein 1; oviductal glycoprotein 1, 120kD; oviductal glycoprotein 1, 120kDa; oviductin; oviduct-specific glycoprotein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12659, 5016 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q12889, Q62010 | |
| OVGP1 | |
| An 18 amino acid peptide from near the amino terminus of human OVGP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction