Learn More
Thermo Scientific™ A2 and A5 Large Gel System Replacement End Gates
Product Code. p-4528814
Click to view available options
Type:
End Gates
Unit Size:
Each
Product Code. 12463657
Brand: Thermo Scientific™ A2EG
Only null left
Add to basket
Add to basket
Use these gasketed UVT gel trays, replacement gaskets and end gates with Thermo Scientific™ A2 and A5 Large Gel Systems.
Use Thermo Scientific™ A2 and A5 Large Gel System Replacement Parts with Owl A2 and A5 Large Gel Systems.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to allow 2, 3 or 4 sample sets to run equal lengths on the same gel
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A2 and A5 Large Gel Electrophoresis System
Type End Gates
Quantity 1 Pair