Learn More
Thermo Scientific™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
Descrizione
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs. These microwell and wall combs are heavy-duty, one-piece units for use with the D3-14, A3-1 and A6 wide gel electrophoresis systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. D3-MTC, -MTD, -MT2C and -MT2D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifica
Specifica
| Description | Microwell comb |
| For Use With (Equipment) | Owl™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System |
| Type | Comb |
| Teeth Count | 50 (2X) |
| Thickness (Metric) Teeth | 1 mm |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.