Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Shop All Thermo Scientific Products
Click to view available options
Type:
Double-sided Comb
Prep Comb
Rapid Load Comb
Standard Comb
Wall Comb
Teeth Count:
1
12
16
2
20
24
25 (2X)
8
9 (1X)
Thickness (Metric) Teeth:
1 mm
1.0/1.5 mm
1.5 mm
Unit Size:
Each
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Ordering Information:
- Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8- and 12-channel pipette format (1X) and (2X) combs
- Customized combs are available (Order SKU#: XCM); please contact your customer service or sales representative for further details
Specifications
Specifications
| For Use With (Equipment) | Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System |
| Gel Size | Mini |
| Product Line | Owl, EasyCast |
| Type | Rapid Load Comb |
| Teeth Count | 9 (1X) |
| Thickness (Metric) Teeth | 1.5 mm |