Learn More
Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts
Product Code. 11530495 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Buffer Drain Bottle
Gaskets
Stopcocks
Temperature Indicators
Unit Size:
Each
Pack of 2
Pack of 5
Product Code. 11530495
Brand: Thermo Scientific™ R10384
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts.
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ S4S Aluminum-Backed Sequencer System
Product Type Buffer Drain Bottle