Learn More
Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Buffer Drain Bottle
Gaskets
Stopcocks
Temperature Indicators
Unit Size:
Each
Pack of 2
Pack of 5
Description
Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.
Choose from these replacement parts for optimal manual sequencing.
Specifications
Specifications
| For Use With (Equipment) | Owl™ S4S Aluminum-Backed Sequencer System |
| Product Type | Stopcocks |
| Quantity | 1 Pair |