missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand (CD252) Mouse anti-Human, BV711, Clone: ik-1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743235
This item is not returnable.
View return policy
Description
OX40 Ligand (CD252) Monoclonal antibody specifically detects OX40 Ligand (CD252) in Human samples. It is validated for Flow Cytometry
Specifications
| OX40 Ligand (CD252) | |
| Monoclonal | |
| Brilliant Violet 711 | |
| CD252; OX40 Ligand; CD134 Ligand; CD134L; TNFSF4; GP34; TXGP1 | |
| Human OX40 Ligand Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| ik-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction