missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (AP-MAB0834) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 1 publication
€ 184.00
Specifications
| Antigen | OX40 Ligand/TNFSF4 |
|---|---|
| Clone | AP-MAB0834 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize |
| Applications | Flow Cytometry |
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| OX40 Ligand/TNFSF4 | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Cell Cycle and Replication, Immunology | |
| PBS with No Preservative | |
| 7292 | |
| Recombinant mouse OX-40 ligand | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
| AP-MAB0834 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| TNFSF4 | |
| IgG2 | |
| Protein A or G purified | |
| CD252 (AP-MAB0834) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title