missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52687JF549
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
Specifications
| OX40Ligand/TNFSF4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TNFSF4 | |
| Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells. | |
| 0.1 mL | |
| Primary | |
| Binds to human OX40L. | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry | |
| R4930 (Oxelumab) | |
| Flow Cytometry | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Rabbit | |
| Protein A or G purified | |
| Cancer, Cell Cycle and Replication, Immunology | |
| 7292 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction