Learn More
Invitrogen™ OX40 Recombinant Rabbit Monoclonal Antibody (125)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560494
Product Code. 30533862
  • € 429.00 / 100µL
Description
OX40 Recombinant Monoclonal Antibody for ELISA
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OX40
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P47741
Tnfrsf4
Recombinant Mouse OX40/TNFRSF4 protein (Met1-Pro211).
100 μL
Primary
Mouse
Antibody
IgG
ELISA
125
Unconjugated
Tnfrsf4
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l
Rabbit
Protein A
RUO
22163
-20°C, Avoid Freeze/Thaw Cycles