Learn More
OX40/TNFRSF4 Antibody (2553B) [DyLight 550], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB10545L
Product Code. 18597495
  • € 605.00 / 0.10mL
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OX40/TNFRSF4
Monoclonal
DyLight 550
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
2553B
Flow Cytometry, CyTOF-ready
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen
Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4, Leu29-Ala216, Accession # P43489
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction
7293
Store at 4°C in the dark.